2-Bromo-α-methylbenzyl alcohol 99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1-(2-Bromophenyl)ethanol
393640250 393640010 393640050
ACRO393640250EA 3250 SEK
ACRO393640250 ACRO393640010 ACRO393640050
2-Bromo-α-methylbenzyl alcohol 99%
2-Bromo-α-methylbenzyl alcohol
Formel: C₈H₉BrO
MW: 201.06 g/mol
Smältpunkt: 54…56 °C
Storage Temperature: Omgivande
MDL Number: MFCD00065001
CAS nummer: 5411-56-3
EINECS: 226-491-3
Order Now
Specifikation testresultat
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5590 to 1.5610 (20°C, 589 nm)
GC ≥98.5 %