Isobuturylklorid 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2-Metylpropanoylklorid
160660010 160660100 160662500
ACRO160660010EA 2825 SEK
ACRO160660010 ACRO160660100 ACRO160662500
Isobuturylklorid 98%
Isobuturylklorid
Formel: C₄H₇ClO
MW: 106,55 g/mol
Smältpunkt: –90 °C
Densitet: 1,017 g/cm³
Flampunkt: 1 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000717
CAS nummer: 79-30-1
EINECS: 201-194-1
UN: 2395
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4060 to 1.4080 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤50 APHA