(±)-2-Aminobutyric acid 99%
Leverantör: Thermo Scientific
Fara
Synonyms: H-DL-Abu-OH, DL-2-Aminobutyric acid, DL-α-Amino-N-Butyric Acid
172091000 172090250
ACRO172091000EA 2225 SEK
ACRO172091000 ACRO172090250
(±)-2-Aminobutyric acid 99%
(±)-2-Aminobutyric acid
Formel: C₄H₉NO₂
MW: 103,12 g/mol
Smältpunkt: 291…293 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00008093
CAS nummer: 2835-81-6
EINECS: 220-616-5
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %
Assay ≥96.0 %