Methyl-4-bromobutyrate 98+%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Methyl 4-bromobutanoate
365830100 365830500
ACRO365830100EA 425 SEK
ACRO365830100 ACRO365830500
Methyl-4-bromobutyrate 98+%
Methyl-4-bromobutyrate
Formel: C₅H₉BrO₂
MW: 181.03 g/mol
Flampunkt: 87 °C
Storage Temperature: Omgivande
MDL Number: MFCD00041482
CAS nummer: 4897-84-1
EINECS: 225-523-3
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4610 to 1.4630 (20°C, 589 nm)
GC ≥98.0 %