1,4-dimetoxibensen ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: p-Dimethoxybenzene, Hydroquinone dimethyl ether
A12822.0E A12822.22 A12822.36
A12822.0EEA 8050 SEK
A12822.0E A12822.22 A12822.36
1,4-dimetoxibensen ≥98%
1,4-dimetoxibensen
Purity: 98% CAS No.: 150-78-7 Molecular Formula: C8H10O2 Formula Weight: 138.17 Flash Point: 94°(201°F) Odour: Sweet Boiling Point: 212° Beilstein Reference: 774605 MDL No.: MFCD00008401
Formel: C₈H₁₀O₂
Kokpunkt: 212 °C
Smältpunkt: 54…58 °C
Densitet: 1,053
Flampunkt: 94 °C (201 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008401
CAS nummer: 150-78-7
EINECS: 205-771-9