7-Azaindole ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 1H-Pyrrolo[2,3-b]pyridine

P1117-25G P1117-5G
TCIAP1117-25GEA 2550 SEK
TCIAP1117-25G TCIAP1117-5G
7-Azaindole ≥98.0% (by GC, titration analysis)
7-Azaindole
Formel: C₇H₆N₂
MW: 118,14 g/mol
Kokpunkt: 270°C
Smältpunkt: 103…106 °C
Densitet: 1,301 g/cm³ (20 °C)
MDL Number: MFCD00005606
CAS nummer: 271-63-6
EINECS: 205-981-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92f545961cfb7e583d452f5956a353d61f44480b&vid=92f545961cfb7e583d452f5956a353d61f44480b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 104.0 to 107.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR