(R)-(-)-2-Methylpiperazine ≥98.0% (by GC, titration analysis)
Leverantör: TCI
|
Fara
|
M2091-25G
M2091-5G
TCIAM2091-25GEA
4400
SEK
TCIAM2091-25G
TCIAM2091-5G
(R)-(-)-2-Methylpiperazine ≥98.0% (by GC, titration analysis)
(R)-(-)-2-Methylpiperazine
Formel:
C₅H₁₂N₂ Kokpunkt: 153 °C (1013 hPa) Smältpunkt: 89…91 °C |
MDL Number:
MFCD00192317 CAS nummer: 75336-86-6 UN: 2925 ADR: 4.1,III |
|
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 99.0 ee% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 88.0 to 92.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...