Veratrol 99+%
Leverantör: Thermo Scientific
Varning
Synonyms: 1,2-Dimethoxybenzene, Pyrocatechol dimethyl ether
161110010 161111000 161115000
ACRO161110010EA 2200 SEK
ACRO161110010 ACRO161111000 ACRO161115000
Veratrol 99+%
Veratrol
Formel: C₈H₁₀O₂
MW: 138,17 g/mol
Kokpunkt: 207 °C (1013 hPa)
Smältpunkt: 15 °C
Densitet: 1,08 g/cm³
Flampunkt: 87 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008357
CAS nummer: 91-16-7
EINECS: 202-045-3
Merck Index: 12,10089
Order Now
Specifikation testresultat
Appearance Clear liquid or solid
Infrared spectrum Conforms
Refractive index 1.5320 to 1.5360 (20°C, 589 nm)
GC ≥99.0 %
Color scale ≤50 APHA