Guajakol 99+%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2-Metoxifenol
120190010 120192500 120190050
ACRO120190010EA 1675 SEK
ACRO120190010 ACRO120192500 ACRO120190050
Guajakol 99+%
Guajakol
Formel: C₇H₈O₂
MW: 124.14 g/mol
Kokpunkt: 205 °C (760 mmHg)
Smältpunkt: 27…29 °C
Densitet: 1.12 g/cm³
Flampunkt: 82 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002185
CAS nummer: 90-05-1
EINECS: 201-964-7
Merck Index: 13,04568
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid or crystalline mass
Infrared spectrum Conforms
Refractive index 1.5390 to 1.5450 (20°C, 589 nm)
GC ≥99.0 %