Provtagare, PharmaPicker
Leverantör: BURKLE
5346-1000 5346-0010 5346-2000
BURK5346-1000EA 7900 SEK
BURK5346-1000 312-0102 BURK5346-2000
Provtagare, PharmaPicker
Provtagare Provtagare, fasta ämnen
PharmaPicker is designed for sampling very small quantities of highly potent or active powders for analysis. The smallest sample quantities can be collected with the PharmaPicker.
  • Tillgänglig separat eller som ett PharmaPicker-set i en praktisk transportlåda
  • Rostfritt stål
  • Elektropolerade
Leveransinformation: PharmaPicker case includes collection cylinders in both sizes, extension rods of 50 and 100 cm, a handle, volume tips (alternative sample volumes on request), and a transport box.