Coagulation tubes, BD Vacutainer®
Leverantör: BD Medical
367704UK 363048 363093UK 367599UK 367562UK
613-3984EA 0 SEK
613-3984 BDAM363048 BDAM363093UK BDAM367599UK BDAM367562UK
Coagulation tubes, BD Vacutainer®
System för klinisk diagnostik Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
PET/PP or glass.
  • With buffered citrate
  • With paper lable
  • Hemogard™ closure
Citrate tubes