4-Nitroveratrole ≥98%

Leverantör: Thermo Scientific
Fara

Synonyms: 3,4-Dimethoxynitrobenzene

A18264.14 A18264.18 A18264.30
A18264.14EA 1075 SEK
A18264.14 A18264.18 A18264.30
4-Nitroveratrole ≥98%
4-Nitroveratrole

Purity: 98+% CAS No.: 709-09-1 Molecular Formula: C8H9NO4 Formula Weight: 183.16 Melting Point: 95-98° Boiling Point: 221-223°/10mm MDL No.: MFCD00007238

Formel: C₈H₉NO₄
MW: 183,16 g/mol
Kokpunkt: 221…223 °C (10 mmHg)
Smältpunkt: 95…98 °C
Densitet: 1,1888 g/cm³ (132.7 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007238
CAS nummer: 709-09-1
EINECS: 211-906-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR