4-Nitroanisol ≥97%
Leverantör: Thermo Scientific
Varning
A19152.0B A19152.30
A19152.0BEA 7150 SEK
A19152.0B A19152.30
4-Nitroanisol ≥97%
4-Nitroanisol
Purity: 97% CAS No.: 100-17-4 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Flash Point: 130°(266°F) Melting Point: 50-54° Boiling Point: 260° Beilstein Reference: 1865361 Merck Reference: 14,6585 UN No.: 3458 MDL No.: MFCD00007327
Formel: C₇H₇NO₃
MW: 153,14 g/mol
Kokpunkt: 260 °C
Smältpunkt: 50…54 °C
Densitet: 1,233
Flampunkt: 130 °C (266 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00007327
CAS nummer: 100-17-4
EINECS: 202-825-3
UN: 3458
ADR: 6.1,III
Merck Index: 14,6585