2,6-Dinitroanilin ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2,6-Dinitrobenzamin
A12120.06 A12120.14 A12120.22
A12120.06EA 1125 SEK
A12120.06 A12120.14 A12120.22
2,6-Dinitroanilin ≥98%
2,6-Dinitroanilin
Purity: 98% CAS No.: 606-22-4 Molecular Formula: C6H5N3O4 Formula Weight: 183.12 Melting Point: ca 136° dec. Beilstein Reference: 2214886 Merck Reference: 14,3271 UN No.: 1596 MDL No.: MFCD00007148
Formel: C₆H₅N₃O₄
MW: 183,12 g/mol
Smältpunkt: ∼ 136 °C (dec.)
Densitet: 1,629 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007148
CAS nummer: 606-22-4
EINECS: 210-108-1
UN: 1596
ADR: 6.1,II
Merck Index: 14,3271