Anilin 99.8%, ren
Leverantör: Thermo Scientific
Fara
Synonyms: Aminobenzene, Phenylamine
158190010 158190050 158190025 158191000
ACRO158190010EA 895 SEK
ACRO158190010 ACRO158190050 ACRO158190025 ACRO158191000
Anilin 99.8%, ren
Anilin
Formel: C₆H₅NH₂
MW: 93,13 g/mol
Kokpunkt: 181…185 °C (760 mmHg)
Smältpunkt: –6,2 °C
Densitet: 1,0213 g/cm³
Flampunkt: 76 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007629
CAS nummer: 62-53-3
EINECS: 200-539-3
UN: 1547
ADR: 6.1,II
Merck Index: 13,00661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3fc5b5071f5a578057c8a7c1c4855ca964c4d840&vid=3bc76a9811da0ba46a79f56c971d11f13dc46900&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow to orange to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5845 to 1.5875 (20°C, 589 nm)
GC ≥99.75 %
Sulfate (SO4) ≤0.03 %
Calcium (Ca) ≤0.005 %
Sodium (Na) ≤0.02 %
Insoluble matter ≤0.005 %