2,4,6-Triiodophenol ≥98%
Leverantör: Thermo Fisher Scientific
Varning
A17145.06 A17145.14 A17145.22
A17145.06EA 895 SEK
A17145.06 A17145.14 A17145.22
2,4,6-Triiodophenol ≥98%
2,4,6-Triiodophenol
Purity: 98% CAS No.: 609-23-4 Molecular Formula: C6H3I3O Formula Weight: 471.80 Melting Point: 156-160° Beilstein Reference: 2046861
Formel: C₆H₃I₃O
Kokpunkt: 316 °C (1013 hPa)
Smältpunkt: 156…160 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002179
CAS nummer: 609-23-4
EINECS: 210-186-7