4-Iodoaniline 99%
Leverantör: Thermo Scientific
Varning
156610250 156611000 156615000
ACRO156610250EA 870 SEK
ACRO156610250 ACRO156611000 ACRO156615000
4-Iodoaniline 99%
4-Iodoaniline
Formel: C₆H₆IN
MW: 219,02 g/mol
Kokpunkt: 262 °C (1013 hPa)
Smältpunkt: 59…64 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00007848
CAS nummer: 540-37-4
EINECS: 208-743-4
UN: 2811
ADR: 6.1,III
Merck Index: 13,05049
Order Now
Specifikation testresultat
Appearance Beige to grey-brown to dark grey Crystalline powder or needles
Infrared spectrum Conforms
Melting point 61 to 64 °C
GC ≥98.5 %