4-Iodobenzoic acid ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: p-Iodobenzoic acid
A10730.09 A10730.18 A10730.30
A10730.09EA 935 SEK
A10730.09 A10730.18 A10730.30
4-Iodobenzoic acid ≥97%
4-Iodobenzoic acid
Purity: 97% CAS No.: 619-58-9 Molecular Formula: C7H5IO2 Formula Weight: 248.02 Melting Point: ca 265° dec. Beilstein Reference: 1860232
Formel: C₇H₅IO₂
MW: 248,02 g/mol
Kokpunkt: 319 °C (1013 hPa)
Smältpunkt: ∼ 265 °C (dec.)
Densitet: 2,18
Storage Temperature: Omgivande
MDL Number: MFCD00002533
CAS nummer: 619-58-9
EINECS: 210-603-2