2-Iodobenzoic acid ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: o-Iodobenzoic acid
A10563.14 A10563.22 A10563.36
A10563.14EA 615 SEK
A10563.14 A10563.22 A10563.36
2-Iodobenzoic acid ≥98%
2-Iodobenzoic acid
Purity: 98+% CAS No.: 88-67-5 Molecular Formula: C7H5IO2 Formula Weight: 248.02 Melting Point: 160-164° Beilstein Reference: 1861406 Merck Reference: 14,5030 MDL No.: MFCD00002419
Formel: C₇H₅IO₂
MW: 248.02 g/mol
Kokpunkt: 314
Smältpunkt: 160…164 °C
Densitet: 2.250
Storage Temperature: Omgivande
MDL Number: MFCD00002419
CAS nummer: 88-67-5
EINECS: 201-850-7
Merck Index: 13,05052