4-Fluorothiophenol, (max. 7% H₂O) 98+%
Leverantör: Thermo Fisher Scientific
Fara
A17600.06 A17600.14 A17600.22
A17600.06EA 1075 SEK
A17600.06 A17600.14 A17600.22
4-Fluorothiophenol, (max. 7% H₂O) 98+%
4-Fluorothiophenol
Purity: 98+% CAS No.: 371-42-6 Molecular Formula: C6H5FS Formula Weight: 128.17 Flash Point: 57°(135°F) Refractive Index: 1.5500 Odour: Unpleasant Boiling Point: 170-172° Beilstein Reference: 1446350 UN No.: 3071 MDL No.: MFCD00004846
Formel: C₆H₅FS
Kokpunkt: 170…172 °C
Densitet: 1,203
Flampunkt: 57 °C (135 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004846
CAS nummer: 371-42-6
EINECS: 206-737-6
UN: 3071
ADR: 6.1,II