5-Fluoro-2-nitrophenol ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 4-Fluoro-2-hydroxynitrobenzene
A12568.09 A12568.18 A12568.30
A12568.09EA 780 SEK
A12568.09 A12568.18 A12568.30
5-Fluoro-2-nitrophenol ≥98%
5-Fluoro-2-nitrophenol
Purity: 98% CAS No.: 446-36-6 Molecular Formula: C6H4FNO3 Formula Weight: 157.10 Flash Point: 91°(195°F) Melting Point: 30-36° Boiling Point: 260° Beilstein Reference: 1870311 MDL No.: MFCD00007107
Formel: C₆H₄FNO₃
Kokpunkt: 260 °C
Smältpunkt: 30…36 °C
Flampunkt: 91 °C (195 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00007107
CAS nummer: 446-36-6
EINECS: 207-168-6