2-Fluoro-6-nitrophenol ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 6-Fluoro-2-nitrophenol
L08851.03 L08851.06
L08851.03EA 1450 SEK
L08851.03 L08851.06
2-Fluoro-6-nitrophenol ≥98%
2-Fluoro-6-nitrophenol
Purity: 98+% CAS No.: 1526-17-6 Molecular Formula: C6H4FNO3 Formula Weight: 157.10 Melting Point: 91-94° Beilstein Reference: 1870307
Formel: C₆H₄FNO₃
MW: 157,1 g/mol
Smältpunkt: 91…94 °C
Storage Temperature: Omgivande
MDL Number: MFCD00042446
CAS nummer: 1526-17-6
EINECS: 216-199-4