2,3,4,5,6-Pentafluoroaniline ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: Aminopentafluorobenzene, Pentafluoroaniline
A19348.06 A19348.14 A19348.22
A19348.06EA 730 SEK
A19348.06 A19348.14 A19348.22
2,3,4,5,6-Pentafluoroaniline ≥98%
2,3,4,5,6-Pentafluoroaniline
Purity: 98+% CAS No.: 771-60-8 Molecular Formula: C6H2F5N Formula Weight: 183.08 Flash Point: 73°(163°F) Melting Point: 34-37° Boiling Point: 156-157° Beilstein Reference: 1819387 MDL No.: MFCD00007643
Formel: C₆H₂F₅N
MW: 183,08 g/mol
Kokpunkt: 156…157 °C
Smältpunkt: 34…37 °C
Densitet: 1,744
Flampunkt: 73 °C (163 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00007643
CAS nummer: 771-60-8
EINECS: 212-234-2