2-Fluoroaniline ≥99%
Leverantör: Thermo Fisher Scientific
Fara
A12612.14 A12612.22 A12612.36
A12612.14EA 635 SEK
A12612.14 A12612.22 A12612.36
2-Fluoroaniline ≥99%
2-Fluoroaniline
Purity: 99% CAS No.: 348-54-9 Molecular Formula: C6H6FN Formula Weight: 111.12 Flash Point: 60°(140°F) Refractive Index: 1.5420 Odour: Recognizable Melting Point: -29° Boiling Point: 171-173° Beilstein Reference: 1524219 UN No.: 2941 MDL No.: MFCD00007642
Formel: C₆H₆FN
Kokpunkt: 171…173 °C
Smältpunkt: –29 °C
Densitet: 1,152
Flampunkt: 60 °C (140 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00007642
CAS nummer: 348-54-9
EINECS: 206-478-9
UN: 2941
ADR: 6.1,III