2,6-Difluoroaniline ≥98%
Leverantör: Thermo Scientific
Varning
A13020.06 A13020.14 A13020.22
A13020.06EA 870 SEK
A13020.06 A13020.14 A13020.22
2,6-Difluoroaniline ≥98%
2,6-Difluoroaniline
Purity: 98% CAS No.: 5509-65-9 Molecular Formula: C6H5F2N Formula Weight: 129.11 Flash Point: 51°(124°F) Refractive Index: 1.5080 Odour: Characteristic Boiling Point: 152-154° Beilstein Reference: 2802697 UN No.: 1992 MDL No.: MFCD00007655
Formel: C₆H₅F₂N
MW: 129,11 g/mol
Kokpunkt: 152…154 °C
Densitet: 1,275
Flampunkt: 51 °C (124 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00007655
CAS nummer: 5509-65-9
EINECS: 226-853-0
UN: 1992
ADR: 3,III