1-Fluoro-4-iodobenzene ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Fluoroiodobenzene
A12008.09 A12008.18 A12008.30
A12008.09EA 1025 SEK
A12008.09 A12008.18 A12008.30
1-Fluoro-4-iodobenzene ≥99%
1-Fluoro-4-iodobenzene
Purity: 99% CAS No.: 352-34-1 Molecular Formula: C6H4FI Formula Weight: 222.00 Flash Point: 68°(154°F) Refractive Index: 1.5830 Melting Point: -20° Boiling Point: 182-184° Beilstein Reference: 1853970 MDL No.: MFCD00001052
Formel: C₆H₄FI
MW: 222 g/mol
Kokpunkt: 182…184 °C
Smältpunkt: –20 °C
Densitet: 1,925
Flampunkt: 68 °C (154 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001052
CAS nummer: 352-34-1
EINECS: 206-522-7