1-Fluoro-4-iodobenzene 99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 4-Fluoroiodobenzene
204500010 204500050 204500500
ACRO204500010EA 330 SEK
ACRO204500010 ACRO204500050 ACRO204500500
1-Fluoro-4-iodobenzene 99%
1-Fluoro-4-iodobenzene
Formel: C₆H₄FI
Kokpunkt: 182…184 °C (760 mmHg)
Smältpunkt: –20 °C
Densitet: 1,925 g/cm³
Flampunkt: 68 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00001052
CAS nummer: 352-34-1
EINECS: 206-522-7
Order Now
Specifikation testresultat
Appearance Clear yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5820 to 1.5840 (20°C, 589 nm)
GC ≥98.5 %