1-Fluoro-3-iodobenzene ≥99%
Leverantör: Thermo Fisher Scientific
Varning
A11706.14 A11706.22
A11706.14EA 1825 SEK
A11706.14 A11706.22
1-Fluoro-3-iodobenzene ≥99%
1-Fluoro-3-iodobenzene
Purity: 99% CAS No.: 1121-86-4 Molecular Formula: C6H4FI Formula Weight: 222.00 Flash Point: 67°(152°F) Refractive Index: 1.5840 Melting Point: -42° Boiling Point: 77-78°/19mm Beilstein Reference: 2039305 MDL No.: MFCD00001044
Formel: C₆H₄FI
Kokpunkt: 77…78 °C (19 mmHg)
Smältpunkt: –42 °C
Densitet: 1,898
Flampunkt: 67 °C (152 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001044
CAS nummer: 1121-86-4
EINECS: 214-339-9