1-Fluoro-2-iodobenzene ≥99% stabilised
Leverantör: Thermo Fisher Scientific
Varning
Stabilization: Stabilised with copper
A12411.14 A12411.22 A12411.36
A12411.14EA 1125 SEK
A12411.14 A12411.22 A12411.36
1-Fluoro-2-iodobenzene ≥99% stabilised
1-Fluoro-2-iodobenzene
Stabilized with copper
Purity: 99% CAS No.: 348-52-7 Molecular Formula: C6H4FI Formula Weight: 222.00 Flash Point: 71°(159°F) Refractive Index: 1.5900 Melting Point: -43° to -40° Boiling Point: 188-189° Beilstein Reference: 2039770 MDL No.: MFCD00001031
Formel: C₆H₄FI
Kokpunkt: 188…189 °C
Smältpunkt: –43 °C to –40 °C
Densitet: 1,903
Flampunkt: 71 °C (159 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001031
CAS nummer: 348-52-7
EINECS: 206-477-3