2,6-Difluorobenzoic acid ≥98%
Leverantör: Thermo Fisher Scientific
Varning
A14558.14 A14558.22
A14558.14EA 1525 SEK
A14558.14 A14558.22
2,6-Difluorobenzoic acid ≥98%
2,6-Difluorobenzoic acid
Purity: 98+% CAS No.: 385-00-2 Molecular Formula: C7H4F2O2 Formula Weight: 158.11 Odour: Odourless Melting Point: 156-160° Boiling Point: 72-77°/13mm Beilstein Reference: 973774 MDL No.: MFCD00002411
Formel: C₇H₄F₂O₂
MW: 158.1 g/mol
Kokpunkt: 72…77 °C (13 mmHg)
Smältpunkt: 156…160 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002411
CAS nummer: 385-00-2
EINECS: 206-856-3