3,4-Difluorobenzonitrile

Leverantör: Thermo Scientific

TL00656DA TL00656EA
MAYBTL00656DAEA 0 SEK
MAYBTL00656DA MAYBTL00656EA
3,4-Difluorobenzonitrile
3,4-Difluorobenzonitrile
Formel: C₇H₃F₂N
MW: 139,1 g/mol
Kokpunkt: 189 °C (1013 hPa)
Smältpunkt: 51 °C
MDL Number: MFCD00011666
CAS nummer: 64248-62-0
EINECS: 264-751-8
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR