3-Fluorobenzaldehyde 98+%
Leverantör: Thermo Fisher Scientific
Varning
119330100 119330500 119332500
ACRO119330100EA 650 SEK
ACRO119330100 ACRO119330500 ACRO119332500
3-Fluorobenzaldehyde 98+%
3-Fluorobenzaldehyde
Formel: C₇H₅FO
MW: 124.11 g/mol
Kokpunkt: 66…68 °C (20 mmHg)
Densitet: 1.17 g/cm³
Flampunkt: 56 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003348
CAS nummer: 456-48-4
EINECS: 207-266-9
UN: 1989
ADR: 3 III
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5170 to 1.5190 (20°C, 589 nm)
GC ≥98.0 %
NMR (1H-NMR) Consistent with structure