4-Chlorothiophenol 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 1-Chloro-4-mercaptobenzene, 4-Chlorobenzene-1-thiol, 4-Chlorobenzenethiol, p-Chlorothiophenol
154760050 154761000 154765000
ACRO154760050EA 480 SEK
ACRO154760050 ACRO154761000 ACRO154765000
4-Chlorothiophenol 98%
4-Chlorothiophenol
Formel: C₆H₅ClS
MW: 144,62 g/mol
Kokpunkt: 205 °C (1013 hPa)
Smältpunkt: 49…53 °C
Densitet: 1,224 g/cm³ (60 °C)
Flampunkt: 109 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004847
CAS nummer: 106-54-7
EINECS: 203-408-9
UN: 3261
ADR: 8,III
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline solid
Infrared spectrum Conforms
Melting point 49 to 53 °C
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure