4-Chloroanisole 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Chloro-4-methoxybenzene
170151000 170150250
ACRO170151000EA 2425 SEK
ACRO170151000 ACRO170150250
4-Chloroanisole 99%
4-Chloroanisole
Formel: C₇H₇ClO
MW: 142,58 g/mol
Kokpunkt: 198…202 °C (760 mmHg)
Smältpunkt: –18 °C
Densitet: 1,16 g/cm³
Flampunkt: 78 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000616
CAS nummer: 623-12-1
EINECS: 210-772-2
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥98.5 %
Specific optical rotation +220° to +235° (20°C, 589 nm) (c=1, H2O)