1-Klor-4-nitrobenzen ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 4-Chloronitrobenzene
A15396.30 A15396.0B A15396.0I
A15396.30EA 785 SEK
A15396.30 A15396.0B A15396.0I
1-Klor-4-nitrobenzen ≥98%
1-Klor-4-nitrobenzen
Purity: 98+% CAS No.: 100-00-5 Molecular Formula: C6H4ClNO2 Formula Weight: 157.56 Flash Point: 127°(260°F) Melting Point: 82-86° Boiling Point: 238-242° Beilstein Reference: 508691 Merck Reference: 14,2151 UN No.: 1578 MDL No.: MFCD00007285
Formel: C₆H₄ClNO₂
MW: 157.56 g/mol
Kokpunkt: 238…242 °C
Smältpunkt: 82…86 °C
Densitet: 1.515
Flampunkt: 127 °C (260 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00007285
CAS nummer: 100-00-5
EINECS: 202-809-6
UN: 1578
ADR: 6.1 II
Merck Index: 13,02170