5-Klor-2-nitroanilin ≥99.0%
Leverantör: TCI
Fara
C1139-25G C1139-250G
TCIAC1139-25GEA 365 SEK
TCIAC1139-25G TCIAC1139-250G
5-Klor-2-nitroanilin ≥99.0%
5-Klor-2-nitroanilin
Formel: C₆H₅ClN₂O₂
MW: 172.57 g/mol
Kokpunkt: 323
Storage Temperature: Omgivande
MDL Number: MFCD00007776
CAS nummer: 1635-61-6
EINECS: 216-661-5
UN: 2237
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=468c6b6d24b59ad53cb1849896dea97767e105aa&vid=18c48717a2281889dba347d36c9d79cea64a3dec&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 99.0 %
Melting point 127.0 to 129.0 deg-C