3,4-Dikloranilin ≥98.0%
Leverantör: TCI EUROPE
Fara
D0324-25G D0324-500G
TCIAD0324-25GEA 215 SEK
TCIAD0324-25G TCIAD0324-500G
3,4-Dikloranilin ≥98.0%
3,4-Dikloranilin
Formel: C₆H₅Cl₂N
MW: 162,02 g/mol
Kokpunkt: 145°C
Smältpunkt: 70…73 °C
Densitet: 1,567 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007768
CAS nummer: 95-76-1
EINECS: 202-448-4
UN: 3442
ADR: 6.1,II
Merck Index: 14,03054
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8730b733b2cc22a117f526d77770e12061e6449d&vid=9cf202fd9d2ccc6dd9e7f84253d07affe3417e2d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 74.0 deg-C
Solubility in Methanol almost transparency