1-Chloro-4-iodobenzene 99%
Leverantör: Thermo Scientific
Varning
146480250 146481000
ACRO146480250EA 820 SEK
ACRO146480250 ACRO146481000
1-Chloro-4-iodobenzene 99%
1-Chloro-4-iodobenzene
Formel: C₆H₄ClI
MW: 238,45 g/mol
Kokpunkt: 226 °C (1013 hPa)
Smältpunkt: 52…55 °C
Densitet: 1,886 g/cm³ (57 °C)
Flampunkt: 108 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001053
CAS nummer: 637-87-6
EINECS: 211-305-5
Order Now
Specifikation testresultat
Appearance Off-white to light beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 52 to 55 °C
GC ≥98.5 %
Heavy metals (as Pb) ≤10 ppm