2-Chloro-6-fluorobenzaldehyde 95%
Leverantör: Thermo Fisher Scientific
Varning
161510250 161511000 161515000
ACRO161510250EA 355 SEK
ACRO161510250 ACRO161511000 ACRO161515000
2-Chloro-6-fluorobenzaldehyde 95%
2-Chloro-6-fluorobenzaldehyde
Formel: C₇H₄ClFO
MW: 158.56 g/mol
Kokpunkt: 92 °C (10 mmHg)
Smältpunkt: 34…39 °C
Flampunkt: 101 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003306
CAS nummer: 387-45-1
EINECS: 206-860-5
Order Now
Specifikation testresultat
Appearance White to yellow Crystalline solid
Infrared spectrum Conforms
Melting point 34 to 39 °C
GC ≥94 %
Specific optical rotation +113° to +119° (20 °C, 589 nm) (c=1, ethanol)