2-Chloro-5-nitrobenzoic acid ≥98%
Leverantör: Thermo Fisher Scientific
Varning
A13754.06 A13754.22 A13754.36
A13754.06EA 560 SEK
A13754.06 A13754.22 A13754.36
2-Chloro-5-nitrobenzoic acid ≥98%
2-Chloro-5-nitrobenzoic acid
Purity: 98+% CAS No.: 2516-96-3 Molecular Formula: C7H4ClNO4 Formula Weight: 201.57 Flash Point: 100°(212°F) Odour: Odourless Beilstein Reference: 1877474 MDL No.: MFCD00007294
Formel: C₇H₄ClNO₄
Smältpunkt: 166…168 °C
Densitet: 0,4 g/cm³ (20 °C)
Flampunkt: >100 °C (212 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00007294
CAS nummer: 2516-96-3
EINECS: 219-739-7