3-Chlorobenzoic acid ≥99%
Leverantör: Thermo Scientific
Varning
A10876.14 A10876.22 A10876.36
A10876.14EA 615 SEK
A10876.14 A10876.22 A10876.36
3-Chlorobenzoic acid ≥99%
3-Chlorobenzoic acid
Purity: 99% CAS No.: 535-80-8 Molecular Formula: C7H5ClO2 Formula Weight: 156.57 Odour: Odourless Melting Point: 154-160° Boiling Point: 274-276° Beilstein Reference: 907218 Merck Reference: 14,2123 MDL No.: MFCD00002491
Formel: C₇H₅ClO₂
MW: 156,57 g/mol
Kokpunkt: 274…276 °C
Smältpunkt: 154…160 °C
Densitet: 1,496 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002491
CAS nummer: 535-80-8
EINECS: 208-618-4
Merck Index: 14,2123