2,5-Diklorbenzonitril

Leverantör: Thermo Scientific

DP01309EA
MAYBDP01309EAEA 0 SEK
MAYBDP01309EA
2,5-Diklorbenzonitril
2,5-Diklorbenzonitril
Formel: C₇H₃Cl₂N
MW: 172,01 g/mol
Kokpunkt: 269 °C (1013 hPa)
Smältpunkt: 130…132 °C
MDL Number: MFCD00019741
CAS nummer: 21663-61-6
EINECS: 244-518-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR