4-Chloro-2-nitrobenzaldehyde ≥98.0%

Leverantör: TCI EUROPE
Varning


M-Clarity™ quality level = MQ300

C0964-25G C0964-5G
TCIAC0964-25GEA 6650 SEK
TCIAC0964-25G TCIAC0964-5G
4-Chloro-2-nitrobenzaldehyde ≥98.0%
4-Chloro-2-nitrobenzaldehyde
Formel: C₇H₄ClNO₃
MW: 185,57 g/mol
MDL Number: MFCD00056104
CAS nummer: 5551-11-1
EINECS: 226-915-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca22ae4392a872f1be0587b812da39b0166210e1&vid=1647fc23c8c3880dcb85a792292f00710324e729&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 66.0 to 69.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR