5-Chloro-2-nitrobenzaldehyde ≥98.0%

Leverantör: TCI
Varning

C0217-25G C0217-5G
TCIAC0217-25GEA 1850 SEK
TCIAC0217-25G TCIAC0217-5G
5-Chloro-2-nitrobenzaldehyde ≥98.0%
5-Chloro-2-nitrobenzaldehyde
Formel: C₇H₄ClNO₃
Storage Temperature: Omgivande
MDL Number: MFCD00007289
CAS nummer: 6628-86-0
EINECS: 229-614-9
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da62fbe48973081c2d867d273b4e483786ee5c96&vid=7ce44743435b5b081cb208d30400f4f6e9ed4fd0&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 73.0 to 78.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR