4-Klorbenzaldehyd 98%
Leverantör: Thermo Fisher Scientific
Varning
150440010 150441000 150442500
ACRO150440010EA 1825 SEK
ACRO150440010 ACRO150441000 ACRO150442500
4-Klorbenzaldehyd 98%
4-Klorbenzaldehyd
Formel: C₇H₅ClO
MW: 140.57 g/mol
Kokpunkt: 214 °C (760 mmHg)
Smältpunkt: >46 °C
Densitet: 1.196 g/cm³
Flampunkt: 88 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003379
CAS nummer: 104-88-1
EINECS: 203-247-4
UN: 3077
ADR: 9 III
Order Now
Specifikation testresultat
Appearance White to light yellow Crystals or crystalline or fused solid
Infrared spectrum Conforms
Melting point 45 to 49 °C
GC ≥97.5 %
Free acid ≤1.5 % (as 4-Chlorobenzoic acid)
Elemental Analysis 17.45 to 18.90 % N