1,4-Diklorbenzen ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: p-Diklorbenzen
A16290.0E A16290.22 A16290.36
A16290.0EEA 1450 SEK
A16290.0E A16290.22 A16290.36
1,4-Diklorbenzen ≥99%
1,4-Diklorbenzen
Purity: 99+% CAS No.: 106-46-7 Molecular Formula: C6H4Cl2 Formula Weight: 147.00 Flash Point: 65°(149°F) Odour: Strong Boiling Point: 173-174° Beilstein Reference: 1680023 Merck Reference: 14,3057 UN No.: 3077 MDL No.: MFCD00000604
Formel: C₆H₄Cl₂
MW: 147 g/mol
Kokpunkt: 173…174 °C
Smältpunkt: 52…56 °C
Densitet: 1,248
Flampunkt: 65 °C (149 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000604
CAS nummer: 106-46-7
EINECS: 203-400-5
UN: 3077
ADR: 9,III
Merck Index: 14,3057