3,4-Dihydroxybenzoic acid ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: Protocatechuic acid
B24016.14 B24016.22 B24016.36
B24016.14EA 1000 SEK
B24016.14 B24016.22 B24016.36
3,4-Dihydroxybenzoic acid ≥97%
3,4-Dihydroxybenzoic acid
Purity: 97% CAS No.: 99-50-3 Molecular Formula: C7H6O4 Formula Weight: 154.12 Melting Point: ca 202° dec. Beilstein Reference: 1448841 Merck Reference: 14,7894 MDL No.: MFCD00002509
Formel: C₇H₆O₄
MW: 154,12 g/mol
Kokpunkt: 125…135 °C (0,001 mmHg)
Smältpunkt: ∼202 °C (dec.)
Densitet: 1,5415 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002509
CAS nummer: 99-50-3
EINECS: 202-760-0
Merck Index: 14,7894