2,6-Dihydroxybenzoic acid ≥98%
Leverantör: Thermo Scientific
Varning
A14485.14 A14485.22 A14485.36
A14485.14EA 1125 SEK
A14485.14 A14485.22 A14485.36
2,6-Dihydroxybenzoic acid ≥98%
2,6-Dihydroxybenzoic acid
Purity: 98% CAS No.: 303-07-1 Molecular Formula: C7H6O4 Formula Weight: 154.12 Melting Point: ca 164° dec. Beilstein Reference: 2209755
Formel: C₇H₆O₄
MW: 154,12 g/mol
Kokpunkt: 339 °C (1013 hPa)
Smältpunkt: ∼164 °C (dec.)
Densitet: 1,58 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002462
CAS nummer: 303-07-1
EINECS: 206-134-8