2,6-Dihydroxybenzoic acid 97%
Leverantör: Thermo Scientific
Varning
114881000 114885000 114880250
ACRO114881000EA 2500 SEK
ACRO114881000 ACRO114885000 ACRO114880250
2,6-Dihydroxybenzoic acid 97%
2,6-Dihydroxybenzoic acid
Formel: C₇H₆O₄
MW: 154,12 g/mol
Kokpunkt: 339 °C (1013 hPa)
Smältpunkt: 160…165 °C
Densitet: 1,58 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002462
CAS nummer: 303-07-1
EINECS: 206-134-8
Order Now
Specifikation testresultat
Appearance White to light yellow to beige Powder or crystals
Infrared spectrum Conforms
Melting point 160 to 165 °C (Dec)
GC ≥96.0 %
Titration with NaOH 96.0 to 104.0 %