2,5-Dihydroxybenzoic acid, Supelco®
Leverantör: Merck
Synonyms: 2,5-Dihydroxybenzoic Acid, DHB, Hydroquinonecarboxylic acid, Gentisic acid, 2,5-DHBA
M-Clarity™ quality level = MQ100
85707-10MG-F 85707-1G-F 85707-250MG-F
85707-10MG-FEA 253 SEK
85707-10MG-F 85707-1G-F 85707-250MG-F
2,5-Dihydroxybenzoic acid, Supelco®
2,5-Dihydroxybenzoic acid
Formel: C₇H₆O₄
MW: 154.12 g/mol
Kokpunkt: 339
Smältpunkt: 202…206 °C
Densitet: 1.55 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002460
CAS nummer: 490-79-9
EINECS: 207-718-5
Merck Index: 13,04409
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75aeb16e8fe84599250924c49b4d1561ca565950&vid=32c94d66a75405b662386e0d2ec04d98b452c314&prid=web&clid=SAPDAM